Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium acetate hydrate is used as a precursor to produce calcium ions in the development of calcium oxide based mesoporous carbon dioxide sorbent. It is an active component of pyroligneous acid-modified limestone. It is employed in the preparation of magnetite-calcium phosphate core-shell nanoparticles. It serves as a raw material in the preparation of hydroxyapatite-wollastonite composites. Further, it acts as a crosslinking agent in the microfluidic production of alginate hydrogel microbeads. In addition to this, it is used to induce crystallization of proteins.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Peterka, T. R.; Grahek, R.; Hren, J.; Bastarda, A.; Bergles, J.; Urleb, U. Solid state compatibility study and characterization of a novel degradation product of tacrolimus in formulation. J. Pharm. Biomed. Anal. 2015, 110, 67-75.
- Niu, S.; Zhou, Y.; Li, H.; Lu, C.; Liu, L. An investigation on the catalytic capability of the modified white mud after activation in transesterification and kinetic calculation. Energy 2015, 89, 982-989.