Thermo Scientific Chemicals

1-Bromo-3,3,3-trifluoroacetone, 97%, Thermo Scientific Chemicals

Catalog number: A14948.22
100 g, Each
Thermo Scientific Chemicals

1-Bromo-3,3,3-trifluoroacetone, 97%, Thermo Scientific Chemicals

Catalog number: A14948.22
100 g, Each
Quantity
Catalog number: A14948.22
also known as A14948-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
431-35-6
IUPAC Name
3-bromo-1,1,1-trifluoropropan-2-one
Molecular Formula
C3H2BrF3O
InChI Key
ONZQYZKCUHFORE-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)CBr
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.3730-1.3780 @ 20?C

Description

1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3,3,3-trifluoroacetone is used in the preparation of 3-nonylthio-1,1,1-trifluoropropan-2-one and perfluoroalkylated trans-allylic alcohols. It is involved in the synthesis of cyclic tetrapeptides. Further, it reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to produce ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text