Thermo Scientific Chemicals

BICINE, 99%

Catalog number: A14957.30
250 g, Each
Thermo Scientific Chemicals

BICINE, 99%

Catalog number: A14957.30
250 g, Each
Quantity
Catalog number: A14957.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
150-25-4
IUPAC Name
sodium 2-[bis(2-hydroxyethyl)amino]acetate
Molecular Formula
C6H12NNaO4
InChI Key
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
SMILES
[Na+].OCCN(CCO)CC([O-])=O
Form
Crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text