Thermo Scientific Chemicals

Tetraethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: A14965.AE
100 mL, Each
Thermo Scientific Chemicals

Tetraethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: A14965.AE
100 mL, Each
Quantity
Catalog number: A14965.AE
also known as A14965-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-10-4
IUPAC Name
tetraethyl silicate
Molecular Formula
C8H20O4Si
InChI Key
BOTDANWDWHJENH-UHFFFAOYSA-N
SMILES
CCO[Si](OCC)(OCC)OCC
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.3810-1.3850 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as cross linking agents in silicon polymers and in aerogel preparations.

Solubility
Soluble in ethanol and 2-propanol.

Notes
Degrades in water. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text