Thermo Scientific Chemicals

4-Chlorophenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A14966.22
100 g, Each
Thermo Scientific Chemicals

4-Chlorophenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A14966.22
100 g, Each
Quantity
Catalog number: A14966.22
also known as A14966-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-12-1
IUPAC Name
1-chloro-4-isocyanatobenzene
Molecular Formula
C7H4ClNO
InChI Key
ADAKRBAJFHTIEW-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)N=C=O
Comment
May develop some turbidity or precipitate
Melting Point
29-33?C
Appearance (Color)
White to cream to yellow
Form
Crystalline or fused solid
Assay (GC)
≥97.5%

Description

4-Chlorophenyl isocyanate is used in the preparation of isothiocyanates. Aryl isocyanates undergo [2+2] cycloaddition reactions with dihydrofuran to give bicyclic β-lactams in high yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenyl isocyanate is used in the preparation of isothiocyanates. Aryl isocyanates undergo [2+2] cycloaddition reactions with dihydrofuran to give bicyclic ß-lactams in high yield.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text