Thermo Scientific Chemicals

(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals

Catalog number: A14967.06
5 g, Each
Thermo Scientific Chemicals

(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals

Catalog number: A14967.06
5 g, Each
Quantity
Catalog number: A14967.06
also known as A14967-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10373-78-1
IUPAC Name
1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Molecular Formula
C10H14O2
InChI Key
VNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILES
CC1(C)C2CCC1(C)C(=O)C2=O
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
199-206?C
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

Solubility
Soluble in ethanol, ethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text