2-Fluorophenol, 98%, Thermo Scientific Chemicals
2-Fluorophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluorophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14979.14
also known as A14979-14
Price (USD)/ Each
63.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
63.60
Add to cart
2-Fluorophenol, 98%, Thermo Scientific Chemicals
Catalog numberA14979.14
Price (USD)/ Each
63.60
-
Add to cart
Chemical Identifiers
CAS367-12-4
IUPAC Name2-fluorophenol
Molecular FormulaC6H5FO
InChI KeyHFHFGHLXUCOHLN-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow to pale brown
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5130-1.5170 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only