Thermo Scientific Chemicals

5-Iodoisatin, 97%, Thermo Scientific Chemicals

Catalog number: A14988.06
5 g, Each
Thermo Scientific Chemicals

5-Iodoisatin, 97%, Thermo Scientific Chemicals

Catalog number: A14988.06
5 g, Each
Quantity
Catalog number: A14988.06
also known as A14988-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20780-76-1
IUPAC Name
5-iodo-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C8H4INO2
InChI Key
OEUGDMOJQQLVAZ-UHFFFAOYSA-N
SMILES
IC1=CC=C2NC(=O)C(=O)C2=C1
Appearance (Color)
Red
Identification (FTIR)
Conforms
Assay (HPLC)
≥96.0%
Form
Powder

Description

5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodoisatin undergoes condensation reaction with phenol yields 5-iodophenolisatin, malonic acid yields 6-iodo-2-quinolone-4-carboxylic acid.

Solubility
Soluble acetone, acetic acid, ethanol. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text