Thermo Scientific Chemicals

1-Dodecene, 96%, Thermo Scientific Chemicals

Catalog number: A14992.36
500 g, Each
Thermo Scientific Chemicals

1-Dodecene, 96%, Thermo Scientific Chemicals

Catalog number: A14992.36
500 g, Each
Quantity
Catalog number: A14992.36
also known as A14992-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-41-4
IUPAC Name
dodec-1-ene
Molecular Formula
C12H24
InChI Key
CRSBERNSMYQZNG-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC=C
Refractive Index
1.4270-1.4330 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥95.0%

Description

1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

Solubility
It is soluble in ethanol, ethyl ether, and acetone

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text