Thermo Scientific Chemicals

Calcium propionate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A14994.0E
2500 g, Each
Thermo Scientific Chemicals

Calcium propionate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A14994.0E
2500 g, Each
Quantity
Catalog number: A14994.0E
also known as A14994-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4075-81-4
IUPAC Name
calcium dipropanoate
Molecular Formula
C6H10CaO4
InChI Key
BCZXFFBUYPCTSJ-UHFFFAOYSA-L
SMILES
[Ca++].CCC([O-])=O.CCC([O-])=O
Appearance (Color)
White
Loss on Drying
≤16.5% (105?C, 2hrs)
Form
Crystals or powder or crystalline powder
Complexometric Titration
≥96.0 to ≤104.0% (dry wt. basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text