Thermo Scientific Chemicals

Bromocyclopropane, 99%, Thermo Scientific Chemicals

Catalog number: A14996.09
10 g, Each
Thermo Scientific Chemicals

Bromocyclopropane, 99%, Thermo Scientific Chemicals

Catalog number: A14996.09
10 g, Each
Quantity
Catalog number: A14996.09
also known as A14996-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4333-56-6
IUPAC Name
bromocyclopropane
Molecular Formula
C3H5Br
InChI Key
LKXYJYDRLBPHRS-UHFFFAOYSA-N
SMILES
BrC1CC1
Form
liquid
Refractive Index
1.4585-1.4615
Appearance (Color)
Clear, colourless to pale yellow
Assay (GC)
>98.5%

Description

Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromo Cyclopropane is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It is also used in the production of cyclopropyl boric acid. It is used as an intermediate in organic synethesis.

Solubility
It is soluble in chloroform, and methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text