Thermo Scientific Chemicals

4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A14999.22
100 g, Each
Thermo Scientific Chemicals

4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals

Catalog number: A14999.22
100 g, Each
Quantity
Catalog number: A14999.22
also known as A14999-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2393-23-9
IUPAC Name
1-(4-methoxyphenyl)methanamine
Molecular Formula
C8H11NO
InChI Key
IDPURXSQCKYKIJ-UHFFFAOYSA-N
SMILES
COC1=CC=C(CN)C=C1
Assay (GC)
≥98.0%
Appearance (Color)
Clear or slightly turbid colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.5440-1.5480 @ 20?C

Description

4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas. Keep away from Strong oxidizing agentsacids, Acid chlorides, Acid anhydrides, Oxidizing agents, Chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text