Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.
Solubility
Soluble in water, methanol, liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.
Notes
Hygroscopic. Incompatible with silver nitrate and strong oxidizers.
RUO – Research Use Only
General References:
- Gao, D.; Chen, C.; Ma, J.; Duan, X.; Zhang, J. Preparation, characterization and antibacterial functionalization of cotton fabric using dimethyl diallyl ammonium chloride-allyl glycidyl ether-methacrylicano-ZnO composite. Chem. Eng. J. 2014, 258, 85-92.
- Chen, H.; Li, G.; Li, H. Novel pretreatment of steam explosion associated with ammonium chloride preimpregnation. Bioresource Technol. 2014, 153, 154-159.