Thermo Scientific Chemicals

Ammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15000.30
250 g, Each
Thermo Scientific Chemicals

Ammonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15000.30
250 g, Each
Quantity
Catalog number: A15000.30
also known as A15000-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-02-9
IUPAC Name
amine hydrochloride
Molecular Formula
ClH4N
InChI Key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES
N.Cl
Form
Crystalline powder
Identification (FTIR)
Conforms
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)
≤0.5%
Appearance (Color)
White

Description

Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.

Solubility
Soluble in water, methanol, liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.

Notes
Hygroscopic. Incompatible with silver nitrate and strong oxidizers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text