Thermo Scientific Chemicals

Ethyl levulinate, 98%, Thermo Scientific Chemicals

Catalog number: A15001.22
100 g, Each
Thermo Scientific Chemicals

Ethyl levulinate, 98%, Thermo Scientific Chemicals

Catalog number: A15001.22
100 g, Each
Quantity
Catalog number: A15001.22
also known as A15001-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
539-88-8
IUPAC Name
ethyl 4-oxopentanoate
Molecular Formula
C7H12O3
InChI Key
GMEONFUTDYJSNV-UHFFFAOYSA-N
SMILES
CCOC(=O)CCC(C)=O
Form
Liquid
Refractive Index
1.4200-1.4240 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text