Thermo Scientific Chemicals

2-Pyridinemethanol, 98+%, Thermo Scientific Chemicals

Catalog number: A15005.22
100 g, Each
Thermo Scientific Chemicals

2-Pyridinemethanol, 98+%, Thermo Scientific Chemicals

Catalog number: A15005.22
100 g, Each
Quantity
Catalog number: A15005.22
also known as A15005-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-98-1
IUPAC Name
(pyridin-2-yl)methanol
Molecular Formula
C6H7NO
InChI Key
SHNUBALDGXWUJI-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=N1
Assay (GC)
≥98.0%
Form
Liquid
Refractive Index
1.5410-1.5450 @ 20?C
Appearance (Color)
Clear colorless to yellow to green to brown

Description

2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyridinemethanol is used as a pyridylalcohol with hypoglycemic activity.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text