Thermo Scientific™

Jump In™ T-REx™ CHO-K1 Kit

Catalog number: A15007
Have Questions?
Thermo Scientific™

Jump In™ T-REx™ CHO-K1 Kit

Catalog number: A15007
Have Questions?
Catalog Number
A15007
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A150071 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Jump-In™ T-REx™ CHO-K1 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ CHO-K1 cell line, to create an inducible isogenic stable cell line with less effort and in less time than traditional cell engineering methods. These cells stably express the tetracycline repressor protein, allowing for inducible expression of your retargeted gene of interest upon the addition of doxycycline.

The high retargeting efficiency, made possible by the R4 sites in the CHO-K1 cell line, allows the use of the isogenic pool for additional experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for the easy selection of a positive stable clone expressing your gene of interest.

The Jump-In™ T-REx™ CHO-K1 Kit lets you:

• Quickly and efficiently develop stably engineered isogenic cell pools in about half the time compared to traditional cell engineering methods
• Inducibly express your gene of interest using doxycycline
• Utilize isogenic expression from a defined genomic locus as the ideal solution for comparative analysis of gene families, isoforms, or orthologs
• Generate multiple cell lines in parallel using the simplified work flow
• Easily access the technology without complicated licenses or restrictions to interpret

Save Time with Rapid and Efficient Generation of Engineered Cell Lines
With the Jump-In™ T-REx™ CHO-K1 Kit you can generate functional cell pools in as little as 2 weeks without laborious clone isolation and analysis, and the streamlined workflow makes it easier to generate several cell lines at the same time. Even generation of clonal cell lines can be done with reduced time and effort due to the high percentage of positive clones. In addition, the Jump-In™ technology gives you the freedom to generate an unlimited number of cell lines without restrictive licensing requirements.

Expand Your Experimental Capabilities
The Jump-In™ T-REx™ CHO-K1 Kit allows for inducible expression of your toxic or unstable gene of interest and is the ideal solution for targets and assays where transient engineering technologies are problematic. The kit also provides a convenient way to create target panels of gene families, isoforms, or orthologs. Genes coding for large proteins or multi-unit proteins are not a problem since the Gateway™ destination vectors accept large inserts.

The kit includes:

Jump-In™ T-REx™ CHO-K1 Cells (2 vials @ 1 ml each)
pJTI™ R4 Dest CMV TO pA Vector (100 μg)--A Gateway™ Destination vector that drives inducible expression of your gene of interest under control of the strong CMV promoter by inclusion of the Tet-Operon.
pJTI™ R4 Int Vector (100 μg)--Expresses the R4 integrase to facilitate the retargeting of your gene of interest into the R4 site.

Sold separately:

pJTI™ R4 CMV-TO MCS pA Vector--A multi-site cloning vector that drives inducible expression of your gene of interest under control of the strong CMV promoter by inclusion of the Tet-Operon.
pJTI™ R4 EXP CMV-TO EmGFP pA Vector--A control vector with inducible expression of GFP under control of the strong CMV promoter by inclusion of the Tet-Operon.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
CHO-K1
Delivery Type
Transfection
Format
Kit
Promoter
CMV/TO
Vector
pJTI™ R4 DEST CMV TO pA , pJTI™ R4 Int
Expression System
Mammalian
For Use With (Application)
Assay development
Inducing Agent
Tetracycline
Key Functions
Stable Cell Line Development, Targeted Integration, Regulated Expression
Product Line
Jump-In™
Product Type
CHO-K1 Kit
Quantity
1 Kit
Selection Agent (Eukaryotic)
Geneticin™ (G-418), Hygromycin, Blasticidin

Contents & Storage

Jump-In™ T-REx™ CHO-K1 Cells (2 vials @ 1 ml each) , store in liquid nitrogen
pJTI™ R4 DEST CMV TO pA vector (100 μg), store at -20 °C
pJTI™ R4 Int vector (100 μg), store at -20 °C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text