It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
Manabu Node; Kiyoharu Nishide; Midori Sai; Kaoru Fuji; Eiichi Fujita. Hard acid and soft nucleophile systems. 3. Dealkylation of esters with aluminum halide-thiol and aluminum halide-sulfide systems. J. Org. Chem. 1981, 46 (10), 1991-1993.
Shea A. Godwin; James R. Cox; Sterling N. Wright. Modeling of benzocaine analog interactions with the D4S6 segment of NaV4.1 voltage-gated sodium channels. Biophysical Chemistry. 2005, 113 (1), 1-7.