Thermo Scientific Chemicals

L-(+)-Asparagine monohydrate, 98+%

Catalog number: A15012.0E
2500 g, Each
Thermo Scientific Chemicals

L-(+)-Asparagine monohydrate, 98+%

Catalog number: A15012.0E
2500 g, Each
Quantity
Catalog number: A15012.0E
also known as A15012-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5794-13-8
IUPAC Name
(2S)-2-amino-3-carbamoylpropanoic acid hydrate
Molecular Formula
C4H10N2O4
InChI Key
RBMGJIZCEWRQES-DKWTVANSSA-N
SMILES
O.N[C@@H](CC(N)=O)C(O)=O
Form
Crystals or powder or crystalline powder or granules
Assay from Supplier's CofA
≥98.0% (U.S. specification)
Optical Rotation
29.5 ± 1.5° (C=5 in 5M HCl, non-U.S. specification)
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0% (non-U.S. specification)

Description

L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text