Thermo Scientific Chemicals

Trimethylacetaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A15013.06
5 g, Each
Thermo Scientific Chemicals

Trimethylacetaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: A15013.06
5 g, Each
Quantity
Catalog number: A15013.06
also known as A15013-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
630-19-3
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Refractive Index
1.3760-1.3820 @ 20°C

Description

Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetaldehyde is used as a stereoselective synthesis application. It is also used as a building block used in aldol condensation reactions. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is virtually insoluble in water but soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from oxidizing agents, strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text