4-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
4-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A15018.36
also known as A15018-36
Price (USD)/ Each
573.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
573.00
Add to cart
4-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Catalog numberA15018.36
Price (USD)/ Each
573.00
-
Add to cart
Chemical Identifiers
CAS156-38-7
IUPAC Name2-(4-hydroxyphenyl)acetic acid
Molecular FormulaC8H8O3
InChI KeyXQXPVVBIMDBYFF-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to pale brown or pale pink
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.

Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. W A Hareland; R L Crawford; P J Chapman; and S Dagley. Metabolic function and properties of 4-hydroxyphenylacetic acid 1-hydroxylase from Pseudomonas acidovorans. J. Bacteriol. 1975, 121 (1), 272-285.
  2. S. Gracin and A. Fischer. 4-Hydroxy­phenyl­acetic acid. Acta Cryst. 2005, 61 o1919-o1920.