Thermo Scientific Chemicals

3,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15038.14
25 g, Each
Thermo Scientific Chemicals

3,4,5-Trimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15038.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15038.14
also known as A15038-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-81-7
IUPAC Name
3,4,5-trimethoxybenzaldehyde
Molecular Formula
C10H12O4
InChI Key
OPHQOIGEOHXOGX-UHFFFAOYSA-N
SMILES
COC1=CC(C=O)=CC(OC)=C1OC
Assay (GC)
≥97.5%
Free acid (titration)
≤1%
Identification (FTIR)
Conforms
Form
Powder or fused solid or flakes
Appearance (Color)
White to cream to pale orange

Description

3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text