3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.
Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
D.V Rao: F.A. Stuber. An efficient synthesis of 3, 4, 5-trimethoxybenzaldehyde from vanillin. Synthesis, 2013, 4 680-689.
Percy S. Manchand; Peter S. Belica; Harry S. Wonga. Synthesis of 3,4,5-Trimethoxybenzaldehyde. Talanta. 1990, 20(17),2659-2666.