Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium n-octadecyl sulfate is used in the formulation of hair care products including shampoo, skin cleansers, and other skin care products.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
Mingins J; and Pethica BA. Properties of ionized monolayers. Part 5.-Surface potentials and pressures of insoluble monolayers of sodium octadecyl sulphate. Faraday Soc., 1963, 591892-1905.
Miriam Gochin; and Lifeng Cai. The role of amphiphilicity and negative charge in glycoprotein 41 interactions in the hydrophobic pocket. Journal of medicinal and pharmaceutical chemistry. 2009, 52(14), 4338-4344.