Thermo Scientific Chemicals

Ethyl pyruvate, 98%, Thermo Scientific Chemicals

Catalog number: A15045.18
50 g, Each
Thermo Scientific Chemicals

Ethyl pyruvate, 98%, Thermo Scientific Chemicals

Catalog number: A15045.18
50 g, Each
Quantity
Catalog number: A15045.18
also known as A15045-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-35-6
IUPAC Name
ethyl 2-oxopropanoate
Molecular Formula
C5H8O3
InChI Key
XXRCUYVCPSWGCC-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4035-1.4075 @ 20?C
Assay (GC)
≥97.5%

Description

Ethyl pyruvate is used as a flavoring agent in food and anti-inflammatory agent for the treatment of critical inflammatory conditions. It is used in the treatment of critical illnesses such as severe sepsis, acute respiratory distress syndrome, burn injury, acute pancreatitis and stroke. It plays an important role in cardiac function after coronary ischemia and reperfusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ethanol and ether.

Notes
Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text