Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.
Solubility
Soluble in water, chloroform, glycerol, glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether.
Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
Shundrina, I. K.; Bukhtoyarova, A. D.; Russkikh, V. V.; Parkhomenko, D. A.; Shelkovnikov, V. V. Synthesis and properties of novel random copolymers made from N-acryloyl piperazine-based monomers and fluoroalkylmethacrylates. Polym. Bull. 2015, 72 (11), 2783-2796.
Zhao, D.; Yu, S.; Liu, G.; Yuan, Q.; Guo, H. Polypiperazine-amide nanofiltration membrane incorporated with poly (ethylene glycol) derivative for electrodialysis concentrate treatment. Sep. Purif. Technol. 2015, 153, 43-50.