Thermo Scientific™

Piperazine, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: A15049.36
500 g, Each
Thermo Scientific™

Piperazine, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: A15049.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15049.36
also known as A15049-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-85-0
Form
Crystals or powder or crystalline powder or fused solid or flakes
Identification (FTIR)
Conforms
Appearance (Color)
White to pale yellow
Assay (GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤1%

Description

Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.

Solubility
Soluble in water, chloroform, glycerol, glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether.

Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text