Thermo Scientific Chemicals

(S)-(+)-Mandelic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A15061.14
25 g, Each
Thermo Scientific Chemicals

(S)-(+)-Mandelic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A15061.14
25 g, Each
Quantity
Catalog number: A15061.14
also known as A15061-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17199-29-0
IUPAC Name
(2S)-2-hydroxy-2-phenylacetic acid
Molecular Formula
C8H8O3
InChI Key
IWYDHOAUDWTVEP-ZETCQYMHSA-N
SMILES
O[C@H](C(O)=O)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥99.0 to ≤101.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
129.0-136.0?C

Description

(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text