Thermo Scientific Chemicals

2-Bromobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15065.36
500 g, Each
Thermo Scientific Chemicals

2-Bromobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15065.36
500 g, Each
Quantity
Catalog number: A15065.36
also known as A15065-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6630-33-7
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%
Free acid (titration)
≤2.0%
Refractive Index
1.5940-1.5980 @ 20?C

Description

2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromobenzaldehyde is used in L-threonine aldolase-catalyzed enantio and diastereoselective aldol reactions. Further, it reacts with trichloromethane to prepare 1-(2-bromo-phenyl)-2,2,2-trichloro-ethanol.

Solubility
Soluble in alcohol and benzene. Insoluble in water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text