Thermo Scientific Chemicals

3-Bromobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: A15075.18
50 g, Each
Thermo Scientific Chemicals

3-Bromobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog number: A15075.18
50 g, Each
Quantity
Catalog number: A15075.18
also known as A15075-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
823-78-9
IUPAC Name
1-bromo-3-(bromomethyl)benzene
Molecular Formula
C7H6Br2
InChI Key
ZPCJPJQUVRIILS-UHFFFAOYSA-N
SMILES
BrCC1=CC=CC(Br)=C1
Appearance (Color)
White
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder or lumps or fused solid
Melting Point (clear melt)
38.0-45.0?C

Description

3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text