3-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals
3-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A15090.22
also known as A15090-22
Price (USD)/ Each
622.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
622.00
Add to cart
3-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Catalog numberA15090.22
Price (USD)/ Each
622.00
-
Add to cart
Chemical Identifiers
CAS3125-64-2
IUPAC Name1-isothiocyanato-3-methoxybenzene
Molecular FormulaC8H7NOS
InChI KeyWHBYCPUKGYEYFU-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6370-1.6410 @ 20?C
Assay (GC)>97.5%
Appearance (Color)Clear colorless to yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only