Thermo Scientific Chemicals

4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15091.22
100 g, Each
Thermo Scientific Chemicals

4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15091.22
100 g, Each
Quantity
Catalog number: A15091.22
also known as A15091-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-35-5
IUPAC Name
4-hydroxybenzene-1,2-dicarboxylic acid
Molecular Formula
C8H6O5
InChI Key
MWRVRCAFWBBXTL-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C=C1C(O)=O
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥97.5%

Description

4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyphthalic acid is used as a precursor to prepare 3,5-dichloro-4-hydroxyphthalic acid using the reagent aqueous acetic acid and chlorine. Further, it is also used as a starting material in the manufacture of N-(4-carboxyphenyl)-4-acetoxyphthalimide.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text