Thermo Scientific Chemicals

4-Hydroxybenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15096.30
250 g, Each
Thermo Scientific Chemicals

4-Hydroxybenzophenone, 98%, Thermo Scientific Chemicals

Catalog number: A15096.30
250 g, Each
Quantity
Catalog number: A15096.30
also known as A15096-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1137-42-4
Appearance (Color)
White to cream to brown or red/brown
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
131-137?C

Description

4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzophenone is used in the field of organic synthesis. It is used as pharmaceutical intermediate of clomifene citrate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text