Thermo Scientific Chemicals

3,5-Dimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15109.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A15109.06
5 g, Each
Quantity
Catalog number: A15109.06
also known as A15109-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7311-34-4
IUPAC Name
3,5-dimethoxybenzaldehyde
Molecular Formula
C9H10O3
InChI Key
VFZRZRDOXPRTSC-UHFFFAOYSA-N
SMILES
COC1=CC(C=O)=CC(OC)=C1
Form
Powder or crystalline solid
Assay (GC)
≥97.5%
Free acid (titration)
<1.0%
Melting Point (clear melt)
44-49?C
Appearance (Color)
White to cream or pale yellow

Description

3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text