Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%, Thermo Scientific Chemicals

Catalog number: A15118.06
5 g, Each
Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%, Thermo Scientific Chemicals

Catalog number: A15118.06
5 g, Each
Quantity
Catalog number: A15118.06
also known as A15118-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1118-71-4
IUPAC Name
2,2,6,6-tetramethylheptane-3,5-dione
Molecular Formula
C11H20O2
InChI Key
YRAJNWYBUCUFBD-UHFFFAOYSA-N
SMILES
CC(C)(C)C(=O)CC(=O)C(C)(C)C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Color (APHA)
≤300 (UK-sourced material)
Assay (Silylated GC)
≥97.5% (UK-sourced material)
Assay from Supplier's CofA
≥97.5% (US-sourced material)

Description

2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text