Thermo Scientific Chemicals

2-Phenylethyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A15123.14
25 g, Each
Thermo Scientific Chemicals

2-Phenylethyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog number: A15123.14
25 g, Each
Quantity
Catalog number: A15123.14
also known as A15123-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2257-09-2
IUPAC Name
(2-isothiocyanatoethyl)benzene
Molecular Formula
C9H9NS
InChI Key
IZJDOKYDEWTZSO-UHFFFAOYSA-N
SMILES
S=C=NCCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5870-1.5910 @ 20?C

Description

Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

Solubility
Soluble in chloroform, ethyl acetate, and dichoromethane. Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text