Thermo Scientific Chemicals

4-Bromoaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A15126.18
50 g, Each
Thermo Scientific Chemicals

4-Bromoaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A15126.18
50 g, Each
Quantity
Catalog number: A15126.18
also known as A15126-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-40-1
IUPAC Name
4-bromoaniline
Molecular Formula
C6H6BrN
InChI Key
WDFQBORIUYODSI-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C=C1
Appearance (Color)
Pale cream to cream to yellow to brown to gray to black
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
59.0-65.0?C

Description

4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

Solubility
Soluble in alcohol and ether. Insoluble in cold water.

Notes
Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text