Thermo Scientific Chemicals

3,5-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A15127.14
25 g, Each
Thermo Scientific Chemicals

3,5-Dichlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A15127.14
25 g, Each
Quantity
Catalog number: A15127.14
also known as A15127-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63352-99-8
IUPAC Name
hydrogen (3,5-dichlorophenyl)hydrazine chloride
Molecular Formula
C6H7Cl3N2
InChI Key
BDXZOJVMTJOAPS-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC(Cl)=CC(Cl)=C1
Appearance (Color)
White to cream
Form
Powder
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%

Description

3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text