Thermo Scientific Chemicals

2'-Fluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: A15128.14
25 g, Each
Thermo Scientific Chemicals

2'-Fluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: A15128.14
25 g, Each
Quantity
Catalog number: A15128.14
also known as A15128-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
445-27-2
IUPAC Name
1-(2-fluorophenyl)ethan-1-one
Molecular Formula
C8H7FO
InChI Key
QMATYTFXDIWACW-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC=C1F
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.5060-1.5110 @ 20?C
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

2'-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoroacetophenone is used as starting reagent in the synthesis of ascididemin. It is used to produce 1-(2-piperidin-1-yl-phenyl)-ethanone by reaction with piperidine.

Solubility
Soluble in Acetone, Chloroform, Dichloromethane, Ethanol, Ethyl Acetate and Methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text