Thermo Scientific Chemicals

2,4-Dichloropyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: A15131.30
250 g, Each
Thermo Scientific Chemicals

2,4-Dichloropyrimidine, 98+%, Thermo Scientific Chemicals

Catalog number: A15131.30
250 g, Each
Quantity
Catalog number: A15131.30
also known as A15131-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3934-20-1
IUPAC Name
2,4-dichloropyrimidine
Molecular Formula
C4H2Cl2N2
InChI Key
BTTNYQZNBZNDOR-UHFFFAOYSA-N
SMILES
ClC1=CC=NC(Cl)=N1
Appearance (Color)
White to yellow
Form
Crystals or crystalline powder and/or chunks
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
57-62?C

Description

2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text