Thermo Scientific Chemicals

Hexamethyldisilazane, 98+%, Thermo Scientific Chemicals

Catalog number: A15139.AP
500 mL, Each
Thermo Scientific Chemicals

Hexamethyldisilazane, 98+%, Thermo Scientific Chemicals

Catalog number: A15139.AP
500 mL, Each
Quantity
Catalog number: A15139.AP
also known as A15139-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
999-97-3
IUPAC Name
bis(trimethylsilyl)amine
Molecular Formula
C6H19NSi2
InChI Key
FFUAGWLWBBFQJT-UHFFFAOYSA-N
SMILES
C[Si](C)(C)N[Si](C)(C)C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4065-1.4105 @ 20?C

Description

Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisilazane is used as a solvent in organic synthesis and organometallic chemistry. It is often used as an adhesion promoter for photoresist in photolithography. Further, it is used for the preparation of trimethylsilyl ethers from hydroxy compounds. It is used as an alternative to critical point drying during sample preparation in electron microscopy. It is added to analyte to get silylated diagnostic products during pyrolysis in gas chromatography- mass spectrometry.

Solubility
Miscible with methanol, chloroform, ethyl ether and benzene. Immiscible with water.                    

Notes
Incompatible with strong oxidizing agents and strong acids.  
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text