Thermo Scientific Chemicals

Propiophenone, 99%, Thermo Scientific Chemicals

Catalog number: A15140.22
100 g, Each
Thermo Scientific Chemicals

Propiophenone, 99%, Thermo Scientific Chemicals

Catalog number: A15140.22
100 g, Each
Quantity
Catalog number: A15140.22
also known as A15140-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
93-55-0
IUPAC Name
1-phenylpropan-1-one
Molecular Formula
C9H10O
InChI Key
KRIOVPPHQSLHCZ-UHFFFAOYSA-N
SMILES
CCC(=O)C1=CC=CC=C1
Form
Liquid
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥98.5%
Refractive Index
1.5245-1.5280 @ 20?C

Description

Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiophenone is an intermediate in the synthesis of pharmaceuticals and organic compounds. It is used in the synthesis of ephedrine and propiophenone derivatives such as cathinone, and methcathinone. It can also be used in the synthesis of aryl alkenes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text