Thermo Scientific Chemicals

(N,N-Dimethyl)methyleneammonium iodide, 97%, Thermo Scientific Chemicals

Catalog number: A15146.18
50 g, Each
Thermo Scientific Chemicals

(N,N-Dimethyl)methyleneammonium iodide, 97%, Thermo Scientific Chemicals

Catalog number: A15146.18
50 g, Each
Quantity
Catalog number: A15146.18
also known as A15146-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33797-51-2
IUPAC Name
(dimethylamino)methylium iodide
Molecular Formula
C3H8IN
InChI Key
VVDUZZGYBOWDSQ-UHFFFAOYSA-M
SMILES
[I-].CN(C)[CH2+]
Appearance (Color)
Pale yellow to yellow or pale orange or cream to brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Iodide)
≥96.0 to ≤104.0%

Description

(N,N-Dimethyl)methyleneammonium iodide is a useful reagent for many synthetic applications like aminomethylation. It is also essential for the conversion of ketones to α, β-unsaturated enones. It is utilized to prepare derivatives of the type RCH2N(CH3)2. It is extensively used in medicines like painkillers and sedatives- Tramadol hydrochloride intermediate viz. 2-dimethylamine methylcyclohexanone. It is also employed in reactive dyes, synthesized spices. It plays an essential role in Mannich reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(N,N-Dimethyl)methyleneammonium iodide is a useful reagent for many synthetic applications like aminomethylation. It is also essential for the conversion of ketones to α, β-unsaturated enones. It is utilized to prepare derivatives of the type RCH2N(CH3)2. It is extensively used in medicines like painkillers and sedatives- Tramadol hydrochloride intermediate viz. 2-dimethylamine methylcyclohexanone. It is also employed in reactive dyes, synthesized spices. It plays an essential role in Mannich reactions.

Solubility
Soluble in dimethylformamide. Partially soluble in acetonitrile, tetrahydrofuran and dichloromethane.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text