Thermo Scientific Chemicals

1-Bromohexadecane, 98%, Thermo Scientific Chemicals

Catalog number: A15147.0B
1000 g, Each
Thermo Scientific Chemicals

1-Bromohexadecane, 98%, Thermo Scientific Chemicals

Catalog number: A15147.0B
1000 g, Each
Quantity
Catalog number: A15147.0B
also known as A15147-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-82-3
Appearance (Color)
Colorless to white to brown
Assay (GC)
≥97.5%
Refractive Index
1.4605-1.4645 @ 20?C
Form
Fused solid or clear liquid as melt
Identification (FTIR)
Conforms

Description

1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexadecane is used in the preparation of soluble carbon nano-onions by covalent functionalization with hexadecyl chains, as extraction solvent in determination of endocrine-disrupting phenols (EDPs) in water samples by ultrasound-assisted emulsification microextraction method.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text