Thermo Scientific Chemicals

Diphenyliodonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15149.18
50 g, Each
Thermo Scientific Chemicals

Diphenyliodonium chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A15149.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15149.18
also known as A15149-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1483-72-3
Appearance (Color)
White to cream
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder

Description

Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.

Solubility
Soluble in water and methanol.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text