Thermo Scientific Chemicals

Methyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A15155.30
250 g, Each
Thermo Scientific Chemicals

Methyl 3-nitrobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: A15155.30
250 g, Each
Quantity
Catalog number: A15155.30
also known as A15155-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-95-1
IUPAC Name
methyl 3-nitrobenzoate
Molecular Formula
C8H7NO4
InChI Key
AXLYJLKKPUICKV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥98.0%
Form
Crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
76-80?C

Description

Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text