Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid disodium salt dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A15161.0I
5000 g, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid disodium salt dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A15161.0I
5000 g, Each
Quantity
Catalog number: A15161.0I
also known as A15161-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6381-92-6
IUPAC Name
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Formula
C10H18N2Na2O10
InChI Key
OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Complexometric Titration
≥99.0 to ≤101.0% (on dried substance)
Appearance (Color)
White
Loss on Drying
8.80-10.55% (1.8-2.2 waters, 200?C/constant wt.)

Description

Ethylenediaminetetraacetic acid disodium salt dihydrate is used as a chelator of divalent cations. It inhibits enzymes such as metalloproteases that require divalent cations for activity. It is also used in chelation therapy. It acts as a sequestering agent in shampoos, cleaners and other personal care products and serves as an anticoagulant for blood samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid disodium salt dihydrate is used as a chelator of divalent cations. It inhibits enzymes such as metalloproteases that require divalent cations for activity. It is also used in chelation therapy. It acts as a sequestering agent in shampoos, cleaners and other personal care products and serves as an anticoagulant for blood samples.

Solubility
Soluble in water.

Notes
Hygroscopic.Incompatible with strong oxidizing agents, strong bases, metals and copper.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text