Thermo Scientific Chemicals

Phenylpropiolic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15163.14
25 g, Each
Thermo Scientific Chemicals

Phenylpropiolic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A15163.14
25 g, Each
Quantity
Catalog number: A15163.14
also known as A15163-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-44-5
IUPAC Name
3-phenylprop-2-ynoic acid
Molecular Formula
C9H6O2
InChI Key
XNERWVPQCYSMLC-UHFFFAOYSA-N
SMILES
OC(=O)C#CC1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥98.0%
Melting Point (clear melt)
133.0-140.0?C
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text