1-Bromohexane, 99%, Thermo Scientific Chemicals
1-Bromohexane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromohexane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
10,000 g
Catalog number A15166.36
also known as A15166-36
Price (USD)/ Each
54.70
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
54.70
Add to cart
1-Bromohexane, 99%, Thermo Scientific Chemicals
Catalog numberA15166.36
Price (USD)/ Each
54.70
-
Add to cart
Chemical Identifiers
CAS111-25-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4460-1.4490 @ 20?C
Identification (FTIR)Conforms
Assay (GC)≥98.5%
FormLiquid
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

Solubility
Immiscible with water.

Notes
Keep away from heat and sources of ignition.
RUO – Research Use Only

General References:

  1. Nwosu, O. S.; Schleicher, J. C.; Scurto, A. M. Kinetics and polarity effects in the synthesis of the ionic liquid, 1-hexyl-3-methyl-imidazolium bromide, using compressed CO 2. J. Supercrit. Fluids 2015, 96, 171-179.
  2. Tiago, G.; Restolho, J.; Forte, A.; Colaço, R.; Branco, L. C.; Saramago, B. Novel ionic liquids for interfacial and tribological applications. Colloids Surf. A 2015, 472, 1-8.