Thermo Scientific Chemicals

trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15167.22
100 g, Each
Thermo Scientific Chemicals

trans-4-Hydroxycinnamic acid, 98%, Thermo Scientific Chemicals

Catalog number: A15167.22
100 g, Each
Quantity
Catalog number: A15167.22
also known as A15167-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
501-98-4
IUPAC Name
(2E)-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular Formula
C9H8O3
InChI Key
NGSWKAQJJWESNS-ZZXKWVIFSA-N
SMILES
OC(=O)C=CC1=CC=C(O)C=C1
Form
Powder
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown to green

Description

trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Hydroxycinnamic acid is used as a hydroxycinnamic acid with antioxidant properties. p-Coumaric acid is an aromatic phytochemical for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text